Documents
DWK212173606
DURAN® Super duty Erlenmeyer narrow-neck flasks
flask capacity (250 mL)
Synonym(s):
212173606
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
flat
Sterilization Compatibility
autoclavable compatible
feature
without cap
packaging
case of 10 ea
manufacturer/tradename
DURAN® 212173606
flask capacity
(250 mL)
General description
Higher impact strength, mechanical strength and stability achieved by reinforced rim. Economical due to longer life span and outstanding corrosion protection. White easy-to-read scale with graduation marks to estimate the content. Fired-on, highly durable white ceramic print.
Other Notes
Legal Information
DURAN is a registered trademark of DWK Life Sciences