Skip to Content
Merck
All Photos(2)

Key Documents

DWK211885403

Duran® Bottles, Reagent, Wide Neck

amber bottle (reagent)

Synonym(s):

211885403

Sign Into View Organizational & Contract Pricing

Select a Size

10 EACH
$3,340.00

$3,340.00


Check Cart for Availability


Select a Size

Change View
10 EACH
$3,340.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

$3,340.00


Check Cart for Availability

material

amber bottle (reagent)
flat head glass cap (stopper)
wide-neck

Joint

Joints ST/NS neck (NS 60/46)

packaging

pkg of 10 ea

manufacturer/tradename

DURAN® 211885403

capacity

1000 mL

Looking for similar products? Visit Product Comparison Guide

General description

The classical stoppered reagent bottles are moulded from amber DURAN® borosilicate 3.3 Type 1 glass, and are suitable for the storage of liquid and solid chemicals and reagents. The bottles feature a wide conical ground joint neck and comply with ISO 4796-2. The wider neck is more convenient for handling powders. The glass stopper provides an excellent seal that protects the contents from evaporation, oxidation, and contamination. The bottles have a Retrace Code for manufacturing lot traceability and a marking field in permanent white enamel. The amber glass ensures that light sensitive contents are protected from the effects of photo-degradation. The protection against ultra-violet light meets international USP/EP standards and lasts the lifetime of the bottle.

Application

Preparation of light-sensitive materials, Storage of light-sensitive materials, Transport and shipping of light-sensitive materials, Autoclaving of media and reagents, Culturing vessel, Boro 3.15

Features and Benefits

USP 660 and USP 671 (Spectral Transmission) compliant

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service