DWK1200265
DURAN® wide mouth laboratory bottle
with cap, round glass bottle, bottle capacity (30,000 mL), non-sterile
Synonym(s):
1200265
Select a Size
$2,390.00
Select a Size
About This Item
$2,390.00
Recommended Products
material
round glass bottle
sterility
non-sterile
feature
with cap
packaging
case of 1 ea
manufacturer/tradename
DURAN® 1200265
bottle capacity
(30,000 mL)
fitting
GL 80 cap
GL 80 neck
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Manufactured from DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
- Easy access to the bottle contents due to the wide diameter of the bottle neck
- Special thread design for rapid opening and closing in less than a turn
- Optimum filling and pouring and particularly easy cleaning
- Spoons, spatulas and tweezers can be manipulated with ease
- Even large-volume funnels can be inserted into the bottles without any difficulty
- Stability due to the large base area
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
- Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service