Skip to Content
Merck
All Photos(2)

Documents

D91055

Sigma-Aldrich

Diethylbenzene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(C2H5)2
CAS Number:
Molecular Weight:
134.22
Beilstein:
1903396
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

~4.6 (vs air)

vapor pressure

0.99 mmHg ( 20 °C)

Assay

95%

form

liquid

expl. lim.

5 %

refractive index

n20/D 1.495 (lit.)

bp

180-182 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

SMILES string

CCc1ccc(CC)cc1.CCc2cccc(CC)c2.CCc3ccccc3CC

InChI

1S/3C10H14/c1-3-9-5-7-10(4-2)8-6-9;1-3-9-6-5-7-10(4-2)8-9;1-3-9-7-5-6-8-10(9)4-2/h3*5-8H,3-4H2,1-2H3

InChI key

WFPVGKJITDSUFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diethylbenzene (DEB) can be used to synthesize divinylbenzene (DVB) by dehydrogenation reaction. DVB is used in the production of crosslinked polystyrene. DEB can be also employed as low temperature heat transfer fluid with biphenyls.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characteristic properties of a locally produced paraffinic oil and its suitability as a heat-transfer fluid
Oyekunle LO and Susu AA
Petroleum Science and Technology, 23(11-12), 1499-1509 (2005)
Gas chromatographic detection of impurities in commercial divinylbenzene
Hannah RE, et al.
Analytical Chemistry, 39(3), 358-360 (1967)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service