Skip to Content
Merck
All Photos(1)

Key Documents

D79004

Sigma-Aldrich

1,5-Dicyanopentane

98%

Synonym(s):

Pimelonitrile, Heptanedinitrile, Pentamethylene dicyanide

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$209.00

$209.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$209.00

About This Item

Linear Formula:
NC(CH2)5CN
CAS Number:
Molecular Weight:
122.17
Beilstein:
1742138
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$209.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.441 (lit.)

bp

175-176 °C/14 mmHg (lit.)

density

0.951 g/mL at 25 °C (lit.)

SMILES string

N#CCCCCCC#N

InChI

1S/C7H10N2/c8-6-4-2-1-3-5-7-9/h1-5H2

InChI key

LLEVMYXEJUDBTA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service