Skip to Content
Merck
All Photos(1)

Key Documents

D70007

Sigma-Aldrich

2,5-Dichlorophenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3OH
CAS Number:
Molecular Weight:
163.00
Beilstein:
1907692
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

chunks
powder or crystals

bp

211 °C (lit.)

mp

54-57 °C (lit.)

SMILES string

Oc1cc(Cl)ccc1Cl

InChI

1S/C6H4Cl2O/c7-4-1-2-5(8)6(9)3-4/h1-3,9H

InChI key

RANCECPPZPIPNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Pao-Kuei Hsiao et al.
International archives of occupational and environmental health, 82(9), 1077-1085 (2009-02-28)
To investigate the effects of 1,4-dichlorobenzene (1,4-DCB) on kidney, liver, and hematological functions of workers in insect repellent factories in Taiwan. A cross-sectional study was performed comparing 46 exposed workers and 29 non-exposed workers. Health information was collected using questionnaires
M A Sogorb et al.
Chemico-biological interactions, 87(1-3), 117-125 (1993-06-01)
The organophosphorus (OP) compound O-hexyl-O-2,5-dichlorophenyl phosphoramidate (H-DCP) is hydrolysed in the plasma, liver and brain of hens and rats. We study in hen plasma the effect of tissue and substrate concentrations and of pH on the hydrolysing activity of H-DCP.
N Díaz-Alejo et al.
Journal of chromatography, 622(2), 179-186 (1993-12-22)
O-Hexyl O-2,5-dichlorophenyl phosphoramidate (HDCP) is a chiral organophosphorus compound that undergoes enzymatic hydrolysis in the rat and hen. Studies of the stereospecificity of its biodegradation are necessary to establish HDCP toxicity. To this effect, methods have been developed for the
R H Hill et al.
Archives of environmental health, 50(4), 277-280 (1995-07-01)
p-Dichlorobenzene is used widely in the United States as a room deodorizer, a moth repellent, and a precursor for a polymer. In a previous study of selected children in Arkansas, we found that 96% of the children had detectable urinary
Xiaoyun Ye et al.
Environment international, 35(8), 1160-1163 (2009-08-12)
In humans, the metabolism of environmental phenols may include the formation of conjugated species (e.g., glucuronides and sulfates), but the free species-not the conjugated forms-are considered biologically active. Therefore, information on the concentration of these free species in blood or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service