Skip to Content
Merck
All Photos(1)

Key Documents

D54206

Sigma-Aldrich

Dichloroacetaldehyde diethyl acetal

≥95.0%

Synonym(s):

1,1-Dichloro-2,2-diethoxyethane

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$273.00
100 G
$589.00

$273.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$273.00
100 G
$589.00

About This Item

Linear Formula:
Cl2CHCH(OC2H5)2
CAS Number:
Molecular Weight:
187.06
Beilstein:
1699086
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$273.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95.0%

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

183-184 °C (lit.)

density

1.138 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(OCC)C(Cl)Cl

InChI

1S/C6H12Cl2O2/c1-3-9-6(5(7)8)10-4-2/h5-6H,3-4H2,1-2H3

InChI key

CDHLQZJRWKQATP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service