D41600
5,7-Dibromo-8-hydroxyquinoline
98%
Synonym(s):
5,7-Dibromo-8-quinolinol, Bromoxin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
98%
form
powder
mp
198-200 °C (lit.)
SMILES string
Oc1c(Br)cc(Br)c2cccnc12
InChI
1S/C9H5Br2NO/c10-6-4-7(11)9(13)8-5(6)2-1-3-12-8/h1-4,13H
InChI key
ZDASUJMDVPTNTF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Reagent for copper, iron and other metals.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 20(6), 987-990 (2004-07-02)
A very simple, ultra-sensitive and highly selective non-extractive spectrophotometric method for the determination of trace amount of cadmium using 5,7-dibromo-8-hydroxyquinoline (DBHQ) has been developed. DBHQ reacts in a slightly acidic solution with cadmium to give a deep greenish-yellow chelate, which
Journal of fluorescence, 29(4), 865-875 (2019-07-22)
Heteroleptic Zn (II) complexes containing 8-hydroxy quinoline as preliminary ligand and pyrazolone based derivatives as secondary ligand were synthesized and their structures confirmed by NMR, Mass, FT-IR, UV-vis and Elemental analysis. Theses complexes show good photoluminescence properties in solid and
Revista iberoamericana de micologia, 33(4), 224-229 (2016-07-28)
Scedosporium species are considered emerging agents causing illness in immunocompromised patients. In Chile, only Scedosporium apiospermum, Scedosporium boydii and Lomentospora prolificans haven been reported previously. The study aimed to characterize genetically Scedosporium dehoogii strains from Chilean soil samples, and assessed
Inorganic chemistry, 59(19), 13858-13874 (2020-09-17)
8-Hydroxyquinolines (8HQs) are a family of lipophilic metal ion chelators that have been used in a range of analytical and pharmaceutical applications over the last 100 years. More recently, CQ (clioquinol; 5-chloro-7-iodo-8-hydroxyquinoline) and PBT2 (5,7-dichloro-2-[(dimethylamino)methyl]-8-hydroxyquinoline) have undergone clinical trials for
European journal of medicinal chemistry, 156, 790-799 (2018-07-29)
8-Hydroxyquinolines (HQ), including clioquinol, possess cytotoxic properties and are widely used as ligands for metal-based anticancer drug research. The number and identity of substituents on the HQ can have a profound effect on activity for a variety of inorganic compounds.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service