Skip to Content
Merck
All Photos(1)

Key Documents

D36607

Sigma-Aldrich

Dibenzyl phosphite

technical grade

Synonym(s):

Dibenzyl phosphonate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$72.50
25 G
$199.00
100 G
$464.00

$72.50


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
$72.50
25 G
$199.00
100 G
$464.00

About This Item

Linear Formula:
(C6H5CH2O)2P(O)H
CAS Number:
Molecular Weight:
262.24
Beilstein:
1982794
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$72.50


Available to ship on22 April 2025Details


Request a Bulk Order

grade

technical grade

Quality Level

form

liquid

refractive index

n20/D 1.555 (lit.)

bp

110-120 °C/0.01 mmHg (lit.)

density

1.187 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O=[PH](OCc1ccccc1)OCc2ccccc2

InChI

1S/C14H15O3P/c15-18(16-11-13-7-3-1-4-8-13)17-12-14-9-5-2-6-10-14/h1-10,18H,11-12H2

InChI key

MIBXHGZAARWAGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francis J Osonga et al.
Molecules (Basel, Switzerland), 25(11) (2020-06-13)
Plant-based pathogenic microbes hinder the yield and quality of food production. Plant diseases have caused an increase in food costs due to crop destruction. There is a need to develop novel methods that can target and mitigate pathogenic microbes. This

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service