Skip to Content
Merck
All Photos(1)

Documents

D227

Sigma-Aldrich

Decafluorobiphenyl

99%

Synonym(s):

1,2,3,4,5-Pentafluoro-6-(2,3,4,5,6-pentafluorophenyl)benzene, 2,2′,3,3′,4,4′,5,5′,6,6′-Decafluoro-1,1′-biphenyl, 2,2′,3,3′,4,4′,5,5′,6,6′-Decafluorobiphenyl, Decafluorodiphenyl, Perfluoro-[1,1′-biphenyl], Perfluorobiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F5C6F5
CAS Number:
Molecular Weight:
334.11
Beilstein:
1916978
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

crystals

bp

206 °C (lit.)

mp

68-70 °C (lit.)

SMILES string

Fc1c(F)c(F)c(c(F)c1F)-c2c(F)c(F)c(F)c(F)c2F

InChI

1S/C12F10/c13-3-1(4(14)8(18)11(21)7(3)17)2-5(15)9(19)12(22)10(20)6(2)16

InChI key

ONUFSRWQCKNVSL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julien Le Roux et al.
Journal of environmental sciences (China), 58, 135-145 (2017-08-05)
The reclamation and disinfection of waters impacted by human activities (e.g., wastewater effluent discharges) are of growing interest for various applications but has been associated with the formation of toxic nitrogenous disinfection byproducts (N-DBPs). Monochloramine used as an alternative disinfectant
Jose Ruiz-Jimenez et al.
Journal of chromatography. A, 1616, 460825-460825 (2020-01-12)
Several calibration approaches were evaluated for the quantitation of volatile organic compounds in air using miniaturized exhaustive and non-exhaustive sampling techniques, such as in-tube extraction (ITEX) and solid phase microextraction (SPME) Arrow. Eleven compounds, 2-ethyl-hexanol, hexanal, nonanal, toluene, ethyl-benzene, methyl
Haopeng Wang et al.
The Analyst, 140(24), 8177-8185 (2015-11-10)
The rising importance of organohalogens in environmental, pharmaceutical, and biological applications has drawn attention to analysis of these compounds in recent years. Elemental mass spectrometry (MS) is particularly advantageous in this regard because of its ability to quantify without compound-specific
Qi Zhang et al.
Archives of environmental contamination and toxicology, 69(2), 133-142 (2015-04-04)
In this study, ice and water samples were collected from seven sites along the Baotou section of the Yellow River during winter 2013. Total polycyclic aromatic hydrocarbon (PAH) concentrations in the ice and in the water under the ice ranged

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service