Skip to Content
Merck
All Photos(1)

Key Documents

D214256

Sigma-Aldrich

5,5-Diphenyl-2-thiohydantoin

99%

Synonym(s):

5,5-Diphenyl-2-thioxo-4-imidazolidinone, 5,5-Diphenylimidazolidine-4-one-2-thione, DPTH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12N2OS
CAS Number:
Molecular Weight:
268.33
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

237-239 °C (lit.)

SMILES string

O=C1NC(=S)NC1(c2ccccc2)c3ccccc3

InChI

1S/C15H12N2OS/c18-13-15(17-14(19)16-13,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H,(H2,16,17,18,19)

InChI key

AMDPNECWKZZEBQ-UHFFFAOYSA-N

Gene Information

Application

Reactant for synthesis of:
  • Imidazole derivatives
  • Imidazothiazole and glycocyamidine derivatives for antimicrobial studies
  • Fatty acid amide hydrolase inhibitor templates
  • Anti-cancer agents
  • Hydantoins and thiohydantoins
  • Acyl CoA: cholesterol acyltransferase inhibitors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheng-Kuo Cheng et al.
Vascular pharmacology, 48(2-3), 138-142 (2008-02-26)
Previously, we identified DPTH, an analogue of antiepileptic drug phenytoin (5,5-diphenylhydantoin, DPT), capable of retarding the cell cycle in the human vascular endothelial cells. Our data suggest that DPTH inhibits human umbilical venous endothelial cells (HUVEC) proliferation by increasing the
Naimeh Moshtael Arani et al.
Ultrasonics sonochemistry, 18(2), 640-643 (2010-10-06)
To obtain a rapid, efficient and mild synthesis of 5,5-diphenylhydantoin and 5,5-diphenyl-2-thiohydantoin derivatives, ultrasonic irradiation has been applied to the reaction mixtures containing substituted benzils and urea or thiourea derivatives catalyzed by KOH in DMSO/H(2)O, which allowed us to achieve
Hitoshi Yoshino et al.
Bioorganic & medicinal chemistry, 18(23), 8150-8157 (2010-11-06)
A series of 5,5-dimethylthiohydantoin derivatives were synthesized and evaluated for androgen receptor pure antagonistic activities for the treatment of castration-resistant prostate cancer. Since CH4933468, which we reported previously, had a problem with agonist metabolites, novel thiohydantoin derivatives were identified by
Yuan Liu et al.
Cancer letters, 271(2), 294-305 (2008-07-25)
Previously, we demonstrated that 5,5-diphenyl-2-thiohydantoin (DPTH) exerts an anti-proliferation effect on subcultured human umbilical vein endothelial cells (HUVEC). In the present study, we show that 2(naphthalen-2-ylmethylsulfanyl)-5,5-diphenyl-1,5-dihydro-imidazol-4-one (DPTH-N10), a derivative compound of DPTH, exerts a 5 times stronger inhibition of [3H]thymidine
H Gerber et al.
Endocrinology, 135(6), 2688-2699 (1994-12-01)
Some years ago, we reported that colloid goiters could be produced experimentally in mice and rats by injection of TSH over a few days in the presence of ample iodine supply. This clearly showed that colloid accumulation and intense TSH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service