Skip to Content
Merck
All Photos(1)

Key Documents

D211508

Sigma-Aldrich

2,2-Diphenylpropane

≥96%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$379.00
25 G
$1,350.00

$379.00


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
5 G
$379.00
25 G
$1,350.00

About This Item

Linear Formula:
(CH3)2C(C6H5)2
CAS Number:
Molecular Weight:
196.29
Beilstein:
2041894
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$379.00


Estimated to ship on14 April 2025


Request a Bulk Order

Quality Level

Assay

≥96%

form

liquid

refractive index

n20/D 1.57 (lit.)

bp

282 °C (lit.)

mp

26-28 °C (lit.)

density

0.992 g/mL at 25 °C (lit.)

SMILES string

CC(C)(c1ccccc1)c2ccccc2

InChI

1S/C15H16/c1-15(2,13-9-5-3-6-10-13)14-11-7-4-8-12-14/h3-12H,1-2H3

InChI key

MILSYCKGLDDVLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service