Skip to Content
Merck
All Photos(1)

Key Documents

D201170

Sigma-Aldrich

Dioctyl sulfosuccinate sodium salt

96%

Synonym(s):

AOT, Bis(2-ethylhexyl) sulfosuccinate sodium salt, DOSS, Docusate sodium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(C2H5)CH2O2CCH2CH(SO3Na)CO2CH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
444.56
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

mp

173-179 °C (lit.)

SMILES string

[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O

InChI

1S/C20H38O7S.Na/c1-5-9-11-16(7-3)14-26-19(21)13-18(28(23,24)25)20(22)27-15-17(8-4)12-10-6-2;/h16-18H,5-15H2,1-4H3,(H,23,24,25);/q;+1/p-1

InChI key

APSBXTVYXVQYAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giovanna Longhi et al.
Physical chemistry chemical physics : PCCP, 13(48), 21423-21431 (2011-11-04)
Positively charged supramolecular aggregates formed in vacuo by n AOTNa (sodium bis(2-ethylhexyl)sulfosuccinate) molecules and n(c) additional sodium ions, i.e. [AOT(n)Na(n+n(c))](n(c)), have been investigated by molecular dynamics (MD) simulations for n = 1-20 and n(c) = 0-5. Statistical analysis of physical
Johnson Mathew et al.
Journal of chromatography. A, 1231, 46-51 (2012-03-01)
Dioctyl sulfosuccinate (DOSS) was a major component of the dispersants most used in the 2010 Deepwater Horizon Oil Spill incident response. This analytical method quantifies salt water DOSS concentrations to a reporting limit of 20 μg/L, which was below the
Sonali V Kapse et al.
International journal of pharmaceutics, 429(1-2), 104-112 (2012-03-15)
We disclose a self nanoprecipitating preconcentrate (SNP) of tamoxifen citrate (TMX), which forms TMX loaded polymeric nanoparticles, on dilution with aqueous media. SNP comprised TMX, polymer (Kollidon SR) and surfactant/s dissolved in a pharmaceutically acceptable vehicle. Binary surfactant mixtures of
Gregory N Smith et al.
Langmuir : the ACS journal of surfaces and colloids, 29(10), 3252-3258 (2013-02-16)
The concentration-dependent aggregation of two surfactants, anionic sodium dioctylsulfosuccinate (Aerosol OT or AOT) and nonionic pentaethylene glycol monododecyl ether (C12E5), has been studied in cyclohexane-D12 using small-angle neutron scattering (SANS). A clear monomer-to-aggregate transition has been observed for both surfactants
Marc-Elie Nader et al.
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery, 146(3), 455-460 (2011-11-22)
(1) To evaluate the ototoxic potential of docusate sodium and mineral oil and (2) to compare the cerumenolytic properties of these agents to water and a commercially available product. Prospective animal study. Docusate sodium and mineral oil are being used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service