Skip to Content
Merck
All Photos(1)

Key Documents

D156604

Sigma-Aldrich

Sodium dimethyldithiocarbamate hydrate

98%

Synonym(s):

Dimethyldithiocarbamic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$189.00
100 G
$560.00

$189.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$189.00
100 G
$560.00

About This Item

Linear Formula:
(CH3)2NCSSNa · aq
CAS Number:
Molecular Weight:
143.21 (anhydrous basis)
Beilstein:
3569024
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$189.00


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

Assay

98%

form

powder

mp

120-122 °C (dec.) (lit.)

SMILES string

[H]O[H].CN(C)C(=S)S[Na]

InChI

1S/C3H7NS2.Na.H2O/c1-4(2)3(5)6;;/h1-2H3,(H,5,6);;1H2/q;+1;/p-1

InChI key

RJCVAPZBRKHUSV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Chelating agent

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathryn L Linge et al.
Environmental science. Processes & impacts, 22(3), 653-662 (2020-03-10)
The use of monochloramine as an alternative disinfectant to chlorine in drinking water treatment can lead to increased formation of emerging nitrogenous halogenated disinfection by-products (DBPs), even when the formation of regulated halogenated DBPs has decreased. In this study, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service