Skip to Content
Merck
All Photos(1)

Documents

D153001

Sigma-Aldrich

1,2-Dimethylcyclohexane, mixture of cis and trans

98%

Synonym(s):

Hexahydro-o-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(CH3)2
CAS Number:
Molecular Weight:
112.21
Beilstein:
1900320
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.432 (lit.)

bp

124 °C (lit.)

density

0.778 g/mL at 25 °C (lit.)

SMILES string

CC1CCCCC1C

InChI

1S/C8H16/c1-7-5-3-4-6-8(7)2/h7-8H,3-6H2,1-2H3

InChI key

KVZJLSYJROEPSQ-UHFFFAOYSA-N

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frank-Dieter Kopinke et al.
Environmental science & technology, 45(23), 10013-10019 (2011-11-01)
This study presents a new experimental technique for measuring rates of desorption of organic compounds from dissolved organic matter (DOM) such as humic substances. The method is based on a fast solid-phase extraction of the freely dissolved fraction of a
Acute toxicities to larval rainbow trout of representative compounds detected in Great Lakes fish.
C C Edsall
Bulletin of environmental contamination and toxicology, 46(2), 173-178 (1991-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service