Skip to Content
Merck
All Photos(1)

Key Documents

D119806

Sigma-Aldrich

Maleic hydrazide

ReagentPlus®, 99%

Synonym(s):

1,2-Dihydro-3,6-pyridazinedione, 3,6-Dihydroxypyridazine, 3,6-Pyridazinediol

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$205.00

$205.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
$205.00

About This Item

Empirical Formula (Hill Notation):
C4H4N2O2
CAS Number:
Molecular Weight:
112.09
Beilstein:
112223
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$205.00


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

product line

ReagentPlus®

Assay

99%

form

powder

autoignition temp.

707 °F

mp

299-301 °C (dec.) (lit.)

SMILES string

O=C1NNC(=O)C=C1

InChI

1S/C4H4N2O2/c7-3-1-2-4(8)6-5-3/h1-2H,(H,5,7)(H,6,8)

InChI key

BGRDGMRNKXEXQD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

572.0 °F - closed cup

Flash Point(C)

300 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maleic hydrazide residues in tobacco and their toxicological implications.
S A Meyer et al.
Reviews of environmental contamination and toxicology, 98, 43-60 (1987-01-01)
L Marcano et al.
Environmental research, 94(2), 221-226 (2004-02-06)
Maleic hydrazide (MH) is an herbicide and is a regulator of the growth of buds in vegetables during storage. It is used in agriculture-in despite its known effect as a mutagenic and clastogenic agent. In this research the effect of
D J Pérez et al.
Environmental pollution (Barking, Essex : 1987), 159(1), 281-286 (2010-10-05)
The wetland macrophyte Bidens laevis possesses suitable cytological characteristics for genotoxicity testing. To test its sensitivity as compared to terrestrial plants species currently in use in standardized assays, Methyl Methanesulfonate (MMS), N-ethyl-N-nitrosourea (ENU) and Maleic Hydrazide (HM) were used. On
Jolanta Juchimiuk et al.
Journal of applied genetics, 48(2), 99-106 (2007-05-15)
The present study is a rare example of a detailed characterization of chromosomal aberrations by identification of individual chromosomes (or chromosome arms) involved in their formation in plant cells by using fluorescent in situ hybridization (FISH). In addition, the first
Tomás Gichner
Mutation research, 538(1-2), 171-179 (2003-07-02)
The purpose of this study was to determine if mutagen-induced DNA damage is correlated with the frequency of induced recombination events. The alkylating agents ethyl methanesulphonate (EMS) and N-ethyl-N-nitrosourea (ENU), and the plant growth regulator and herbicide maleic hydrazide (MH)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service