Skip to Content
Merck
All Photos(1)

Key Documents

D103209

Sigma-Aldrich

(+)-Digitoxigenin

98%

Synonym(s):

3β,14-Dihydroxy-5β,20(22)-cardenolide, 3,14,21-Trihydroxy-20(22)-norcholenic acid lactone, 5β,20(22)-Cardenolide-3β,14-diol, Digitoxigenin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H34O4
CAS Number:
Molecular Weight:
374.51
Beilstein:
95448
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

optical activity

[α]20/D +16.9°, c = 1.3 in methanol

mp

253-255 °C (dec.) (lit.)

SMILES string

C[C@]12CC[C@H](O)C[C@H]1CC[C@@H]3[C@@H]2CC[C@]4(C)[C@H](CC[C@]34O)C5=CC(=O)OC5

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Machiko Ono et al.
Chemical & pharmaceutical bulletin, 60(8), 1076-1082 (2012-08-07)
The addition of 4 eq of chloral to osmundalactone (4S,5R)-4 gave quantitative formation of the hemiacetal derivative (4S,5R)-8, which was treated with methane sulfonic acid to afford the intramolecular Micheal addition product (+)-(3S,4S,5R)-9 possessing a 3,4-cis-dihydroxy-δ-lactone in 78% overall yield
Maoquan Zhou et al.
Organic letters, 8(19), 4339-4342 (2006-09-08)
A convergent and stereocontrolled route to trisaccharide natural product digitoxin has been developed. The route is amenable to the preparation of both the digitoxigen mono- and bisdigitoxoside. This route featured the iterative application of the palladium-catalyzed glycosylation reaction, reductive 1,3-transposition
A Babes et al.
Biophysical journal, 79(5), 2557-2571 (2000-10-29)
We have used admittance analysis together with the black lipid membrane technique to analyze electrogenic reactions within the Na(+) branch of the reaction cycle of the Na(+)/K(+)-ATPase. ATP release by flash photolysis of caged ATP induced changes in the admittance
Margarita Ramirez et al.
Biochemical pharmacology, 70(6), 851-857 (2005-08-06)
Compound 14beta,17beta-cycloketoester-3beta-OH androstane (INCICH-D7) is a semisynthetic product of a structural modification of the digitoxigenin molecule. INCICH-D7 has a heterocyclic ketoester type fusion between positions C14 and C17 of the steroid nucleus, which confers this molecule stronger electronegativity than that
Rodrigo M Pádua et al.
Steroids, 73(4), 458-465 (2008-02-06)
A simple and versatile method for the chemical synthesis of 21-hydroxypregnane 21-O-malonyl hemiesters which may be important intermediates of cardenolide biosynthesis is described. Starting from commercial beta-methyldigitoxin, acid hydrolysis followed by 3beta-O-acetylation and ozonolysis with reductive cleavage of the ozonides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service