Skip to Content
Merck
All Photos(1)

Key Documents

CLS9999913

Corning® disposable phenolic cap

13-415 thread

Synonym(s):

phenolic caps, threaded cap, threaded caps

Sign Into View Organizational & Contract Pricing

Select a Size

1000 EA
$255.00

$255.00


Available to ship on28 April 2025Details



Select a Size

Change View
1000 EA
$255.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.43

$255.00


Available to ship on28 April 2025Details


manufacturer/tradename

Corning 99999-13

H

13.5 mm

fitting

13-415 thread

Looking for similar products? Visit Product Comparison Guide

General description

These disposable Corning caps are made from phenolic resin and are autoclavable at 121°C. Caps are provided with a white rubber liner.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service