Key Documents
CLS9990125
Pyrex® ribbed watch glasses
O.D. ~125 mm
Synonym(s):
Corning® 9990
Select a Size
12 EA
$203.00
$203.00
Estimated to ship on23 April 2025
Select a Size
Change View
12 EA
$203.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$203.00
Estimated to ship on23 April 2025
manufacturer/tradename
Corning 9990-125
O.D.
~125 mm
General description
Molded-in ribs allow vapors to escape during boiling to eliminate pressure buildup. Meets EPA requirements for methods 3005, 3010, 3020.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)