CLS9944716
Pyrex® disposable culture tubes, screw cap style with white marking spot
capacity 15.5 mL
Synonym(s):
culture tube, glass tubes, test tubes
Select a Size
$785.00
Select a Size
About This Item
$785.00
Recommended Products
material
borosilicate glass
round bottom tube
sterility
non-sterile
feature
cap: no
packaging
case of 1000
pack of 250
manufacturer/tradename
Corning 99447-16
O.D. × L
16 mm × 125 mm
capacity
15.5 mL
color
clear
fitting
thread for 15-415
suitability
suitable for (culture)
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service