Key Documents
CLS7732
Pyrex® gum stability glass bombs
gum bomb w/ cover, reusable, non-sterile, 1/cs
Synonym(s):
lab equipment accessory
Select a Size
1 EA
$89.70
$89.70
Estimated to ship on01 May 2025
Select a Size
Change View
1 EA
$89.70
About This Item
UNSPSC Code:
41121500
NACRES:
NB.43
$89.70
Estimated to ship on01 May 2025
material
borosilicate glass (borosilicate glass code 7740)
cap (Pourout)
sterility
non-sterile
feature
reusable
packaging
case of 1
pack of 1 ×
manufacturer/tradename
Corning 7732
O.D. × H
50 mm × 105 mm
suitability
suitable for (the determination of gum stability of gasoline)
General description
These PYREX® bombs are for the determination of gum stability of gasoline. The cover prevents condensed vapors in bomb stem from contaminating the sample, but does not interfere with free flow of oxygen. Made in accordance with ASTM D-525.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)