Key Documents
CLS69473LBO
Pyrex® resin reaction kettle bottom
capacity 3,000 mL
Synonym(s):
pyrex kettle, reaction equipment, reaction flasks
Select a Size
1 EA
$841.00
4 EA
$1,680.00
$841.00
Estimated to ship on25 May 2025
Select a Size
Change View
1 EA
$841.00
4 EA
$1,680.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$841.00
Estimated to ship on25 May 2025
material
borosilicate glass
wide-mouth
sterility
non-sterile
feature
closure type lid
packaging
pack of 1
case of 4
manufacturer/tradename
Corning 6947-3LBO
availability
not available in EU
capacity
3,000 mL
General description
Flange is finely ground for a tight seal.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)