CLS5680250
Pyrex® volumetric flask, certified and serialized, with Pyrex® ST stopper
capacity 250 mL
Synonym(s):
flasks glass, volumetric flasks, volumetric glass, Corning® 5680
Select a Size
$70.70
Select a Size
About This Item
$70.70
Recommended Products
material
borosilicate glass
long
stopper size
16
sterility
non-sterile
feature
class A
closure type ST stopper
packaging
case of 1
manufacturer/tradename
Corning 5680-250
tol. measuring range
0.12 mL
capacity
250 mL
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service