CLS55802L
Pyrex® class A volumetric flask with snap cap
capacity 2,000 mL
Synonym(s):
clear flasks, pyrex flask, volumetric flasks
Select a Size
$252.00
Available to ship on28 April 2025Details
Select a Size
About This Item
$252.00
Available to ship on28 April 2025Details
Recommended Products
material
borosilicate glass
long
sterility
non-sterile
feature
class A
closure type Snap cap
packaging
pack of 1
case of 4
manufacturer/tradename
Corning 5580-2L
Corning 5580
tol. measuring range
0.50 mL
capacity
2,000 mL
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service