Key Documents
CLS5400500
Pyrex® iodine determination flask, with Pyrex® ST stopper
capacity 500 mL
Synonym(s):
flask erlenmeyer, flasks erlenmeyer, glass flasks, Corning® 5400
Select a Size
6 EA
$392.00
$392.00
Check Cart for Availability
Select a Size
Change View
6 EA
$392.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$392.00
Check Cart for Availability
material
borosilicate glass
wide-mouth
Joint
joint ST/NS stopper22
sterility
non-sterile
feature
cap: no
closure type ST stopper
packaging
case of 6
manufacturer/tradename
Corning 5400-500
capacity
500 mL
General description
Flasks are blown in specillay designed molds, thereby insuring uniformity and proper contour. The stopper projects above the liquid seal trough to facilitate removal.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)