Key Documents
CLS502050
Pyrex® narrow-mouth Erlenmeyer flask with ST stopper
capacity 50 mL
Synonym(s):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, Corning® 5020
Select a Size
12 EA
$290.00
$290.00
Check Cart for Availability
Select a Size
Change View
12 EA
$290.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$290.00
Check Cart for Availability
material
borosilicate glass
bottom (baffled)
glass cap
narrow-mouth
Joint
joint stopper19
sterility
non-sterile
feature
cap
closure type Stopper
packaging
case of 12
manufacturer/tradename
Corning 5020-50
capacity
50 mL
Other Notes
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)