Key Documents
CLS49505L
Pyrex® round-bottom vertical three neck distilling flask, with 24/40 joints
capacity 5,000 mL
Synonym(s):
bottom flask, flask round, flask round bottom, Corning® 4950
Select a Size
3 EA
$1,350.00
$1,350.00
Estimated to ship on14 April 2025
Select a Size
Change View
3 EA
$1,350.00
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
$1,350.00
Estimated to ship on14 April 2025
material
borosilicate glass
round bottom flask
three-neck (Vertical)
Joint
joint ST/NS24/40
sterility
non-sterile
feature
closure type Standard taper joint
packaging
pack of 1
case of 3
manufacturer/tradename
Corning 4950-5L
capacity
5,000 mL
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)