CLS44464L
Pyrex® shaker flask with extra-deep baffles
capacity 4,000 mL
Synonym(s):
Corning® 4446
Select a Size
$794.00
Select a Size
About This Item
$794.00
Recommended Products
material
Delong
borosilicate glass
sterility
non-sterile
feature
cap: no
closure type Closure cap
packaging
case of 2
manufacturer/tradename
Corning 4446-4L
capacity
4,000 mL
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)