Key Documents
CLS40606L
Pyrex® Florence flat-bottom flask
capacity 6,000 mL
Synonym(s):
bottom flask, flat flask, florence flask, Corning® 4060
Select a Size
1 EA
$161.00
$161.00
Check Cart for Availability
Select a Size
Change View
1 EA
$161.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$161.00
Check Cart for Availability
material
borosilicate glass
flat bottom flask
stopper size
11
sterility
non-sterile
feature
closure type Tooled mouth
long neck, tooled mouth
packaging
case of 1
manufacturer/tradename
Corning 4060-6L
capacity
6,000 mL
neck size
(1)
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)