Skip to Content
Merck
All Photos(1)

Key Documents

CLS3838

Corning® 1536 well plates

flat bottom clear polystyrene wells, low base, TC-treated, sterile, lid

Synonym(s):

1536 multiwell plate, 1536 well microplate, 1536 well microtiter plate, 1536 well plate, HTS plate, TC-treated plate, immunoassay plate, tissue-culture treated plate

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$1,150.00
50 EA
$3,460.00

$1,150.00


Check Cart for Availability


Select a Size

Change View
10 EA
$1,150.00
50 EA
$3,460.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
black plate
flat bottom clear polystyrene wells
polystyrene plate
size:
1536 wells

$1,150.00


Check Cart for Availability

material

black plate
flat bottom clear polystyrene wells
polystyrene plate

sterility

sterile

feature

lid
skirt
plate format: low base, with lid

packaging

pkg of 10 ea
case of 50 ea

manufacturer/tradename

Corning 3838

size

1536 wells

well volume

12.8 μL
up to 8 μL

color

clear

suitability

suitable for (fluorescent; luminescent assays)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Corning 1536 Well Plates

  • 1536 well polystyrene plates
  • Low background fluorescence, minimal light scatter and reduced crosstalk
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Opaque walls to prevent well-to-well crosstalk
  • Flat well bottoms possess superior flatness -minimizes variability within a plate and between plates
  • Round well bottoms for reduced air entrapment and improved CV′s and Z factor
  • Raised well bottoms for high sensitivity
  • Flood reservior on four sides to reduce instrument contamination
  • Suitable for fluorescent and luminescent assays

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service