CLS3581
Corning® disposable spinner flasks
with vent caps, capacity 3 L, sterile
Synonym(s):
cell culture flasks, cell culture spinner flasks, scale up cell culture flasks
Select a Size
$629.00
Select a Size
About This Item
$629.00
Recommended Products
material
high-density polyethylene cap
polystyrene flask
vent cap
sterility
sterile
sterile
feature
cap
closure type Vented cap
packaging
case of 4
manufacturer/tradename
Corning 3581
capacity
3 L
working volume
1-3 L
1000-3000 mL
fitting
cap for GL 45
GL 45 neck
suitability
suitable for (scale-up cell culture; production of secreted proteins)
General description
Convenience
The Corning disposable spinner flask system comes ready-to-use with paddle and integrated magnet, eliminating the need for time-consuming assembly or cleaning and reassembly.
Confidence
Molded from virgin polystyrene and gamma-irradiated, each spinner flask system assures a clean and sterile unit that is nonpyrogenic. No more concerns with detergent residues or contamination.
Compatibility
Made of ISO 10993 polystyrene, the vessel is comparable to conventional glass spinner flasks for growth of suspension cell lines and any attachment-dependent cultures using microcarrier beads.
Performance
The paddle size and height is optimized for each vessel size. A unique integrated magnet provides a smooth, even rotation at required speeds on any laboratory slow speed stirrer. Heat build-up in the vessel is reduced by means of a specially designed flange that raises the vessel off the stir-plate surface.
Conclusions
- Disposable large volume spinner flasks are equivalent to conventional glass spinner flasks for cell growth and viability
- A variety of cell types can be cultured in large volume disposable spinner flasks
- Disposable spinner flasks maintain equal aeration as compared to standard glass spinner flasks
- Cultured cells from glass and disposable spinner flasks are metabolically equivalent
- Culured cells in disposable spinner flasks product equal amounts of secreted proteins as cells cultured in traditional glass spinner flasks
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)