CLS30621L
Pyrex® class A serialized/certified single metric scale graduated cylinder
volume 1,000 mL
Synonym(s):
Corning® 3062
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
subdivision 10.0 mL
manufacturer/tradename
Corning 3062-1L
tol. measuring range
3.0 mL
volume
1,000 mL
Looking for similar products? Visit Product Comparison Guide
General description
Calibrated "to deliver".
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service