CLS2962300
Pyrex® hydrometer jar
capacity 300 mL
Synonym(s):
Corning® 2962
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Corning 2962-300
O.D. × H
39 mm × 381 mm
capacity
300 mL
General description
Breakage is minimized by the heavy wall construction. The large hexagonal base, sealed to the cylinder body, increases stability. Meets ASTM specifications D-287 and E-100.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.