Skip to Content
Merck
All Photos(1)

Key Documents

CDP-502

Sigma-Aldrich

Cationic Liposomes

DOTAP-based - DOTAP:DOPC (40:60 molar ratio 10mM)

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
$993.00

$993.00


Estimated to ship on11 May 2025


Request a Bulk Order

Select a Size

Change View
2 ML
$993.00

About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

$993.00


Estimated to ship on11 May 2025


Request a Bulk Order

Quality Level

composition

Phosphate buffered saline

concentration

10 mM

impurities

40 mol % DOTAP
60 mol % DOPC

particle size

100 nm

pH

7.4

Looking for similar products? Visit Product Comparison Guide

General description

Liposomes are extensively used to study the interaction of proteins, peptides and other molecules with the surface of a lipid membrane. One of the parameters that affects this interaction is the charge of the liposomal membrane. DOTAP is a positively charged molecule that is combined with DOPC as a matrix lipid. Various amounts of cationic DOTAP from 0.5% to 50% results in liposomes with various zeta potentials. Charged liposomes, such as the cationic DOTAP-based liposomes, can be used in various in vitro studies and drug delivery applications. The cationic DOTAP-based liposomes have varying degrees of postive charge and can be used to study the interaction of these liposomes with various types of enzymes, proteins, peptides and other moelcules that interact with the surface of liposomes.

Application

Drug delivery
Lipid-protein interactions

Storage and Stability

Liposomes should never be frozen. Liposomes should be stored in the dark at 4°C, except when brought to room temperature for brief periods prior to use.

Liposomes are made under sterile conditions. If you need to take multiple aliquots out of the vial, it is advised to take extreme care in not contaminating the vial. It is recommended to handle the vial under a sterile hood to maintain the sterility of the product. Liposomes should never be frozen. Ice crystals that form during freezing will rupture the lipid membrane of the liposomes and change the size of liposomes particles.

Legal Information

Cellsome is a trademark of Encapsula NanoSciences
Product of Encapsula Nanosciences

Disclaimer

For research use only

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service