Skip to Content
Merck
All Photos(1)

Key Documents

C80806

Sigma-Aldrich

Cinnamamide, predominantly trans

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$75.80
25 G
$244.00

$75.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$75.80
25 G
$244.00

About This Item

Linear Formula:
C6H5CH=CHCONH2
CAS Number:
Molecular Weight:
147.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$75.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

mp

148-150 °C (lit.)

SMILES string

[H]\C(=C(\[H])c1ccccc1)C(N)=O

InChI

1S/C9H9NO/c10-9(11)7-6-8-4-2-1-3-5-8/h1-7H,(H2,10,11)/b7-6+

InChI key

APEJMQOBVMLION-VOTSOKGWSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastiano Masuri et al.
Metallomics : integrated biometal science, 12(6), 891-901 (2020-04-28)
The novel copper complex [Cu(phen)2(salubrinal)](ClO4)2 (C0SAL) has been synthesised and characterised. Copper(ii) is coordinated by salubrinal through the thionic group, as shown by the UV-Vis, IR, ESI-MS and tandem mass results, together with the theoretical calculations. The formed complex showed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service