Skip to Content
Merck
All Photos(1)

Documents

C41207

Sigma-Aldrich

2-Chloroethyl ethyl ether

99%

Synonym(s):

2-Ethoxyethyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2OCH2CH3
CAS Number:
Molecular Weight:
108.57
Beilstein:
1731361
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.411 (lit.)

bp

107 °C (lit.)

density

0.989 g/mL at 25 °C (lit.)

SMILES string

CCOCCCl

InChI

1S/C4H9ClO/c1-2-6-4-3-5/h2-4H2,1H3

InChI key

GPTVQTPMFOLLOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service