Skip to Content
Merck
All Photos(1)

Key Documents

C117609

Sigma-Aldrich

Cyclopropanecarbonitrile

98%

Synonym(s):

Cyanocyclopropane, Cyclopropyl cyanide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$59.00

$59.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$59.00

About This Item

Linear Formula:
C3H5CN
CAS Number:
Molecular Weight:
67.09
Beilstein:
635790
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$59.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.421 (lit.)

bp

135 °C (lit.)

density

0.911 g/mL at 25 °C (lit.)

SMILES string

N#CC1CC1

InChI

1S/C4H5N/c5-3-4-1-2-4/h4H,1-2H2

InChI key

AUQDITHEDVOTCU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service