Key Documents
BR91214
BRAND® glass beaker with spout, low form
volume 25 mL, with graduation
Synonym(s):
Beakers, Glass
Select a Size
10 EA
$195.00
$195.00
Available to ship on22 April 2025Details
Select a Size
Change View
10 EA
$195.00
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
$195.00
Available to ship on22 April 2025Details
material
borosilicate glass 3.3
feature
with graduation
manufacturer/tradename
BRAND 91214
volume
25 mL
General description
The BRAND® Glass Beaker with Spout is expertly crafted from Boro 3.3 glass, making it a reliable choice for laboratory applications. Designed in accordance with DIN EN ISO 3819 standards, this low-form beaker features clear graduation markings for precise measurement of liquids. The integrated spout allows for easy pouring, minimizing spills and enhancing accuracy during liquid transfer.
Other Notes
Browse all glass beakers
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)