BR782861
BRAND® insert strips for 24 x 6-well BRANDplates®
Inlet Opening System, PC membrane, pore size 8 μm
Synonym(s):
BRANDplates®, Cell culture, Insert Strips, Insert System, Inserts, Tissue culture
Select a Size
$805.00
Select a Size
About This Item
$805.00
Recommended Products
material
PC membrane
description
cellGrade™ plus
sterility
sterile
packaging
pack of 12 ea (strips of 4 inserts, individually wrapped)
manufacturer/tradename
BRAND 782861
size
6 well
pore size
8 μm
binding type
non-treated surface
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)