Skip to Content
Merck
All Photos(1)

Key Documents

BR782746

BRAND® 6-well Cell Culture Insert, PC-membrane

pore size 8.0 μm, pack of 24 pieces

Sign Into View Organizational & Contract Pricing

Select a Size

24 EA
$573.00

$573.00


Check Cart for Availability


Select a Size

Change View
24 EA
$573.00

About This Item

UNSPSC Code:
41122107
NACRES:
NB.13

$573.00


Check Cart for Availability

material

PC membrane

packaging

pack of 24 pieces

manufacturer/tradename

BRAND 782746

size

(6 well)

pore size

8.0 μm

Looking for similar products? Visit Product Comparison Guide

General description

The BRAND Insert 2in1 is a cell culture insert used for skin model production, 3D epithelium cultures, chemotaxis analyses, transmigration assays, transport and polarity studies, and more.

In many cell culture tasks, or when switching from a standing to a hanging position, you can continue to use your current cell culture plates, saving additional validation steps. Thanks to both supporting feet and hooks, you can position the BRAND Insert 2in1 either way.

Via its protein-like structure, the cellGrade plus surface provides optimal conditions for growing primary keratinocytes and many other cell types.

With the practical multipacks, the right number of inserts is always close at hand. They can be opened row by row, so that the remaining inserts stay sterile. Besides the practical benefits, this also reduces waste.


Overview

  • Use in a hanging or standing position
  • Works with all common 6-, 12-, or 24-well multi-well plates
  • Suitable for 2D and 3D cell cultures
  • Surface: cellGrade plus
  • Sterility Assurance Level (SAL) of 10-6
  • PC or PET membrane
  • Pore size: from 0.4 μm and 8.0 μm
  • Packaged individually or in practical multi-packs

Features and Benefits

  • Inserts can be used in a hanging or standing position
  • For 24-, 12-, and 6-well plates or culture dishes
  • Achieve uniform and controlled cell growth thanks to surface treatment, without coatings

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
cellGrade is a trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The BRAND® Insert 2in1 cell culture insert supports the cultivation of Reconstructed Human Epidermis used for skin corrosion tests.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service