BR781961
BRAND® microplate BRANDplates®, cellGrade
size 96 wells, size 330 μL, V-bottom, transparent
Synonym(s):
BRANDplates®, Cell culture, Microplates, Tissue culture, cellGrade
Select a Size
$356.00
Select a Size
About This Item
polystyrene
transparent wells
96 wells
skirt (V-bottom)
with lid
$356.00
Recommended Products
material
V-bottom
polystyrene
transparent wells
sterility
sterile
feature
lid
skirt (V-bottom)
with lid
packaging
pack of 50 ea
pkg of (individually packed)
manufacturer/tradename
BRAND 781961
size
330 μL
96 wells
working volume
330 μL
color
transparent
binding type
cell culture treated
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Optimal surfaces for optimal results
- Reliable quality from the cleanroom
- Tested by independent laboratories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service