BR781842
BRAND® microplate BRANDplates®, lipoGrade
size 96 wells, size 350 μL, flat bottom, transparent
Synonym(s):
BRANDplates®, Immunology, Microplates, lipoGrade
Select a Size
$680.00
Estimated to ship on07 May 2025
Select a Size
About This Item
flat bottom
polystyrene
96 wells
$680.00
Estimated to ship on07 May 2025
Recommended Products
material
colorless wells
flat bottom
polystyrene
sterility
non-sterile
feature
lid: no
skirt (F-bottom)
packaging
pack of 100 ea
manufacturer/tradename
BRAND 781842
size
350 μL
96 wells
working volume
350 μL
color
transparent
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service