BR781721
BRAND® microplate BRANDplates®, immunoGrade
size 96 wells, size 360 μL, V-bottom, transparent
Synonym(s):
BRANDplates®, Immunology, Microplates, high binding, immunoGrade
Select a Size
$404.00
Select a Size
About This Item
colorless wells
polystyrene
96 wells
$404.00
Recommended Products
material
V-bottom
colorless wells
polystyrene
sterility
non-sterile
feature
lid: no
skirt (V-bottom)
packaging
pack of 100 ea
manufacturer/tradename
BRAND 781721
size
360 μL
96 wells
working volume
360 μL
color
transparent
suitability
suitable for (bacteriological assays)
binding type
high binding surface
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Optimal surfaces for optimal results
- Reliable quality from the cleanroom
- Tested by independent laboratories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service