BR759210
BRAND® UV cuvette micro
center H 8.5 mm, volume 70-850 μL, pack of 500 ea
Synonym(s):
Cuvettes, UV-Cuvettes
Select a Size
$293.00
Available to ship on28 April 2025Details
Select a Size
About This Item
$293.00
Available to ship on28 April 2025Details
Recommended Products
packaging
pack of 500 ea
manufacturer/tradename
BRAND 759210
center H
8.5 mm
volume
70-850 μL
General description
- BRAND® UV cuvette micro is manufactured under controlled room conditions.
- Ideal for the determinations in water analysis, chemistry, and life science applications.
- Cuvette material offers high chemical resistance to most polar solvents, acids, and alkaline solutions.
- Clear, clean optical path with an indication of optical path orientation.
- Eliminates the risk of cross-contamination.
- Grouped by mold cavity number.
- An economical alternative to Quartz cuvettes.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)