BR705912
BRAND® Transferpette® S Multichannel Pipette
capacity (30-300 μL), 8-channel, adjustable volume, M8-300, DE-M
Synonym(s):
BRAND Multichannel Pipette, Transferpette S
Select a Size
$1,540.00
Select a Size
About This Item
$1,540.00
Recommended Products
feature
8-channel adjustable volume
packaging
case of 1 ea
manufacturer/tradename
BRAND 705912
capacity
(30-300 μL)
Related Categories
General description
Overview- Volume range of single-channel pipette: from 0.1 μl to 10 ml- Volume range of multi-channel pipette: from 0.5 μl to 300 μl- Short stroke and low operating forces- Universal tip cone- Completely autoclaving at 121 μC- One-hand operation for right- and left-handed users- Color code on volume adjustment wheel, shaft and volume-change protection- Easy insertion and ejection of tips due to O-rings on multi-channel pipettes- Can be calibrated easily and without tools thanks to Easy Calibration technology- Slim shaft for sampling from narrow vessels (centrifuge tubes)- Operating temperature from +15 μC to +40 μC- Vapor pressure up to 500 mbar- Viscosity up to 260 mPa s
Efficient: One-hand operation
Flexible: ergonomic design for all hand sizes
Reliable: volume-change protection prevents accidental adjustments
Features and Benefits
- Efficient: one-hand operation
- Flexible: ergonomic design for all hand sizes
- Reliable: volume-change protection prevents accidental adjustments
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)