BR705828
BRAND® Transferpette® S Pipette
tip volume (50 μL), single-channel, fixed volume, mechanical, DE-M
Select a Size
$408.00
Select a Size
About This Item
$408.00
Recommended Products
feature
single-channel fixed volume
mechanical
packaging
case of 1 ea
manufacturer/tradename
BRAND 705828
tip volume
(50 μL)
Related Categories
Features and Benefits
- Efficient: one-hand operation
- Flexible: ergonomic design for all hand sizes
- Reliable: volume-change protection prevents accidental adjustments
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)