Skip to Content
Merck
All Photos(1)

Key Documents

BR705210

BRAND® HandyStep® Touch S

DE-M, with universal AC adapter and shelf/rack mount

Synonym(s):

Electronic Pipette, HandyStep, HandyStep S, HandyStep Touch, HandyStep Touch S, Positive Displacement Pipettte, Repeater, Repeater Pipette, Repetitive Pipette

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$2,330.00

$2,330.00


Available to ship on24 April 2025Details



Select a Size

Change View
1 EA
$2,330.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.02

$2,330.00


Available to ship on24 April 2025Details


manufacturer/tradename

BRAND 705210

General description

Repetitive pipettes are the first choice when it comes to efficient dispensing of long series. Positive displacement pipettes are often the right solution for pipetting viscous media. The new stepper HandyStep touch from BRAND combines both features.

With the touchscreen display, you conveniently swipe back and forth between functions and always have an overview of the essential information on your work steps. Operation is easy, even with protective gloves on. And speaking of easy, the days of manually changing tips are over. On the HandyStep touch, lightly tapping on the Eject icon on the display is enough to eject the tip.

The HandyStep touch repetitive pipette automatically recognizes the volume of the new BRAND PD- Tips II. This saves time and reduces the risk of error. Tips from many other manufacturers can also be used, providing maximum flexibility.
Charging is easy and efficient: simply place the HandyStep touch in the inductive charging stand and the battery automatically starts charging.
Overview

  • Touchscreen operation with intuitive menu navigation in English, German, Spanish, French and Chinese
  • HandyStep touch with the features multi-dispensing, auto-dispensing and pipetting; HandyStep touch S with the additional features sequential dispensing, multi-aspiration and titration
  • Automatic size detection of BRAND PD-Tips II
  • Compatible with PD-Tips prior to the PD-Tips II generation and with tips from many other manufacturers: for example, Encode tips, Repet-Tips, Combitips, Combitips plus, Combitips advanced (the above-mentioned brands are property of their respective owners)
  • Automatic tip ejection
  • Variable volume adjustment from 1.0 μl to 50 ml
  • Easy to clean thanks to smooth surface without grooves and edges
  • Inductively rechargeable

Application

auto-pipetting, multi-dispensing, sequential dispensing, multi-aspiration, multi-titration

Features and Benefits

Overview:
  • Touchscreen operation with intuitive menu navigation in English, German, Spanish, French and Chinese
  • Multi-dispensing, auto-dispensing and pipetting
  • Automatic size detection of BRAND PD-Tips II
  • Compatible with PD-Tips prior to the PD-Tips II generation and with tips from many other manufacturers: for example, Encode tips, Repet-Tips, Combitips®, Combitips plus®, Combitips advanced® (the above-mentioned brands are property of their respective owners)
  • Automatic tip ejection
  • Variable volume adjustment from 1.0 µl to 50 ml
  • Easy to clean thanks to smooth surface without grooves and edges
  • Inductively rechargeable


Limitations of use:
  • Operating temperature of instrument and reagent should be between +15 °C and +40 °C (59 °F to 104 °F)
  • Vapor pressure up to 500 mbar
  • Viscosity: At 50 ml PD-Tip: 20 mPa s; at 5 ml PD-Tip: 260 mPa s; at 1,25 ml PD-Tip: 977 mPa s

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Combitips is a registered trademark of Eppendorf AG
Combitips advanced is a registered trademark of Eppendorf AG
Combitips plus is a registered trademark of Eppendorf AG
HandyStep is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service