Skip to Content
Merck
All Photos(1)

Key Documents

BR701367

BRAND® sealing film for storage

DMSO-resistant, allows visual inspection

Synonym(s):

Accessories, Deep-well Plates, Plates, Sample Storage, Sealing films

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$218.00

$218.00


Check Cart for Availability


Select a Size

Change View
100 EA
$218.00

About This Item

UNSPSC Code:
41122410
NACRES:
NB.01

$218.00


Check Cart for Availability

material

polypropylene
polypropylene film

sterility

non-sterile

feature

piercable

packaging

pack of 100 ea

manufacturer/tradename

BRAND 701367

General description

This film is ideal for PCR, ELISA and sample storage. Due to its adhesive, it can be used down to - 80° C, is DMSO-resistant and resistant to common solvents.

Features and Benefits

  • Temperature stability up to 120°C
  • Tight seal for minimizing evaporation
  • DMSO- and solvent-resistant

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service