Key Documents
BR55646
BRAND® conical joint clip
polyoxymethylene, for NS 29/32
Synonym(s):
Conical joint stopper
Select a Size
1 EA
$16.40
$16.40
In StockDetails
Select a Size
Change View
1 EA
$16.40
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
$16.40
In StockDetails
material
polyoxymethylene
red
description
Operating temperature up to 150 °C
packaging
pack of 1 ea
manufacturer/tradename
BRAND 55646
General description
Will not scratch glass surfaces. Excellent resilience and stability. Outstanding chemical resistance.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)