BR4600121
BRAND® Dispensette® S Analog-adjustable bottle-top dispenser
volume range 0.2-2 mL, with recirculation valve
Synonym(s):
BRAND Dispensette, Bottle-Top Dispenser
Select a Size
$872.00
Available to ship on28 April 2025Details
Select a Size
About This Item
$872.00
Available to ship on28 April 2025Details
Recommended Products
feature
incremental 0.05 mL
DE-M marking
with recirculation valve
manufacturer/tradename
BRAND 4600121
volume range
0.2-2 mL
fitting
(GL 28/S28)
GL 32
GL 38
S 40
General description
Features and Benefits
- Extremely easy filling and dispensing
- Simple priming by optimized flow channels
- Rapid and safe volume adjustment with internal scalloped track
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
Protocols
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Related Content
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.